Calcium propionate hydrate 97%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Propionic acid calcium salt hydrate
A14994.0E
A14994.36
A14994.0EEA
3030
CZK
A14994.0E
A14994.36
Calcium propionate hydrate 97%
Calcium propionate hydrate
Purity: 97%
CAS No.: 4075-81-4
Molecular Formula: (CH3CH2COO)2Ca.xH2O
Formula Weight: 186.22(anhy)
Melting Point:
Vzorec:
(CH₃CH₂COO)₂Ca·xH₂O MW: 186.22 g/mol (anhy) Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167354 CAS číslo: 4075-81-4 EINECS: 223-795-8 Merck Index: 14,1698 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...