Tisk…

Stearan sodný

Dodavatel: Thermo Fisher Scientific

Synonyms: Stearate sodium, Stearic acid sodium salt

39648.A1 39648.A7 39648.30
39648.A1EA 2930 CZK
39648.A1 39648.A7 39648.30
Stearan sodný
Stearan sodný

CAS No.: 822-16-2 Molecular Formula: NaCO2(CH2)16CH3 Formula Weight: 306.46 Physical Form: Powder Odour: Weak Solubility: Slowly soluble in cold water. Solubility increases with temperature MDL No.: MFCD00036404

Vzorec: C₁₈H₃₅NaO₂
MW: 306.46
Bod tání: 288
Hustota: 1.103 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036404
CAS číslo: 822-16-2
EINECS: 212-490-5
Merck Index: 14,8678

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR