Sodium butyrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Butyric acid sodium salt, Sodium Butyrate
M-Clarity™ quality level = MQ200
303410-100G
303410-500G
303410-5G
SIAL303410-100GEA
1710
CZK
SIAL303410-100G
SIAL303410-500G
SIAL303410-5G
Sodium butyrate, Sigma-Aldrich®
Sodium butyrate
Vzorec:
C₄H₇NaO₂ Bod tání: 244 °C Hustota: 1,324 g/cm³ (30 °C) |
MDL Number:
MFCD00002816 CAS číslo: 156-54-7 EINECS: 205-857-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...