Tisk…

Undecanoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Hendecanoic acid, Undecylic acid

M-Clarity™ quality level = MQ300

W324507-100G W324507-1KG W324507-5KG W324507-SAMPLE
SIALW324507-100GEA 1860 CZK
SIALW324507-100G SIALW324507-1KG SIALW324507-5KG SIALW324507-SAMPLE
Undecanoic acid, Sigma-Aldrich®
Undecanoic acid
Vzorec: C₁₁H₂₂O₂
Bod varu: 228 °C (1013 hPa)
Bod tání: 30 °C
Hustota: 0,89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002730
CAS číslo: 112-37-8
EINECS: 203-964-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR