Tisk…

Pentadecanoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Pentadecylic acidn-Pentadecanoic acid

M-Clarity™ quality level = MQ100

P6125-1G P6125-25G P6125-5G
SIALP6125-1GEA 980 CZK
SIALP6125-1G SIALP6125-25G SIALP6125-5G
Pentadecanoic acid, Sigma-Aldrich®
Pentadecanoic acid
Vzorec: C₁₅H₃₀O₂
MW: 242,4 g/mol
Bod varu: 339 °C (1013 hPa)
Bod tání: 51…53 °C
Hustota: 0,8496 g/cm³ (70 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002745
CAS číslo: 1002-84-2
EINECS: 213-693-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR