Tisk…

Margaric acid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Heptadecanoic acid, Heptadecylic acid

M-Clarity™ quality level = MQ200

H3500-1G H3500-25G H3500-5G
SIALH3500-1GEA 1590 CZK
SIALH3500-1G SIALH3500-25G SIALH3500-5G
Margaric acid, Sigma-Aldrich®
Margaric acid
Vzorec: C₁₇H₃₄O₂
MW: 270,46 g/mol
Bod varu: 364 °C (1013 hPa)
Bod tání: 63 °C
Hustota: 1,02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002751
CAS číslo: 506-12-7
EINECS: 208-027-1
Merck Index: 13,05775

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR