Tisk…

Arachidic acid

Dodavatel: Cayman Chemical

Synonyms: Eicosanylic acid, Arachidinic acid, Icosanoic acid, Eicosanoic acid

9000339-100 9000339-50
CAYM9000339-100EA 1540 CZK
CAYM9000339-100 CAYM9000339-50
Arachidic acid
Arachidic acid
Vzorec: C₂₀H₄₀O₂
MW: 312.54
Bod varu: 328
Bod tání: 75.5
Hustota: 0.838 (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00002755
CAS číslo: 506-30-9
EINECS: 208-031-3
Merck Index: 12,00802

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR