Arachidic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Eicosanylic acid, Arachidinic acid, Icosanoic acid, Eicosanoic acid
A19616.03
A19616.06
A19616.14
A19616.03EA
1040
CZK
A19616.03
A19616.06
A19616.14
Arachidic acid ≥98%
Arachidic acid
Vzorec:
C₂₀H₄₀O₂ MW: 312.54 Bod varu: 328 Bod tání: 74…76 °C Hustota: 0.838 (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00002755 CAS číslo: 506-30-9 EINECS: 208-031-3 Merck Index: 14,764 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...