Tributyl citrate ≥99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Citric acid tributyl ester, Hydroxytricarballylic acid tributyl ester, Tributyl 2-hydroxypropane-1,2,3-tricarboxylate
A10266.0B
A10266.30
A10266.0BEA
4160
CZK
A10266.0B
A10266.30
Tributyl citrate ≥99%
Tributyl citrate
Purity: 99%
CAS No.: 77-94-1
Molecular Formula: C18H32O7
Formula Weight: 360.45
Flash Point: 157°(315°F)
Refractive Index: 1.4440
Odour: Weak
Melting Point: -20°
Boiling Point: 325°
Beilstein Reference: 1806072
Merck Reference: 14,1564
MDL No.: MFCD00027217
Vzorec:
C₁₈H₃₂O₇ Bod varu: 325 °C Bod tání: –20 °C Hustota: 1,040 Bod vzplanutí: 157 °C (315 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00027217 CAS číslo: 77-94-1 EINECS: 201-071-2 Merck Index: 14,1564 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...