Dimethyl adipate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Adipic acid dimethyl ester, 1,6-Hexanedioic acid dimethyl ester
M-Clarity™ quality level = MQ100
186252-100G
186252-500G
SIAL186252-100GEA
1810
CZK
SIAL186252-100G
SIAL186252-500G
Dimethyl adipate, Sigma-Aldrich®
Dimethyl adipate
Vzorec:
C₈H₁₄O₄ Bod varu: ∼229 °C (1013 hPa) Hustota: 1,06 g/cm³ (20 °C) Bod vzplanutí: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008469 CAS číslo: 627-93-0 EINECS: 211-020-6 Merck Index: 13,00163 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...