Tisk…

Diethyladipát ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: NSC 19160, Adipic acid diethyl ester, 1,6-Hexanedioic acid diethyl ester, NSC 3363

A11476.22 A11476.36
A11476.22EA 944 CZK
A11476.22 A11476.36
Diethyladipát ≥99%
Diethyladipát

Purity: 99% CAS No.: 141-28-6 Molecular Formula: C10H18O4 Formula Weight: 202.25 Flash Point: 110°(230°F) Refractive Index: 1.4275 Melting Point: -20° Boiling Point: 250-252° Beilstein Reference: 1780035 Merck Reference: 14,162 MDL No.: MFCD00009215

Vzorec: C₁₀H₁₈O₄
Bod varu: 250…252 °C
Bod tání: –20 °C
Hustota: 1,008
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009215
CAS číslo: 141-28-6
EINECS: 205-477-0
Merck Index: 14,162

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR