Tisk…

Dimethyl glutarate ≥98%

Dodavatel: Thermo Fisher Scientific

B24359.14 B24359.22
B24359.14EA 821 CZK
B24359.14 B24359.22
Dimethyl glutarate ≥98%
Dimethyl glutarate

Purity: 98% CAS No.: 1119-40-0 Molecular Formula: C7H12O4 Formula Weight: 160.17 Flash Point: 103°(217°F) Refractive Index: 1.4240 Melting Point: -37° Boiling Point: 213-215° Beilstein Reference: 1771444 Merck Reference: 14,4473 MDL No.: MFCD00008468

Vzorec: C₇H₁₂O₄
Bod varu: 213…215 °C
Bod tání: –37 °C
Hustota: 1,087
Bod vzplanutí: 103 °C (217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008468
CAS číslo: 1119-40-0
EINECS: 214-277-2
Merck Index: 14,4473

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR