Dimethyl succinate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
DBE-4 dibasic ester, Succinic acid dimethyl ester, 1,4-Butanedioic acid dimethyl ester
M-Clarity™ quality level = MQ300
W239607-10KG-K
W239607-1KG-K
W239607-25KG-K
SIALW239607-10KG-KEA
13560
CZK
SIALW239607-10KG-K
SIALW239607-1KG-K
SIALW239607-25KG-K
Dimethyl succinate, Sigma-Aldrich®
Dimethyl succinate
Vzorec:
C₆H₁₀O₄ MW: 146.14 Bod varu: 200 Bod tání: 18…19 °C Hustota: 1.117 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008466 CAS číslo: 106-65-0 EINECS: 203-419-9 Merck Index: 12,09038 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...