Tisk…

Dimethyl malonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

136441-1KG 136441-250G
SIAL136441-1KGEA 4430 CZK
SIAL136441-1KG SIAL136441-250G
Dimethyl malonate, Sigma-Aldrich®
Dimethyl malonate
Vzorec: C₅H₈O₄
Bod varu: 181 °C (1013 hPa)
Bod tání: –62 °C
Hustota: 1,15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008460
CAS číslo: 108-59-8
EINECS: 203-597-8
Merck Index: 12,06169

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR