Dimethyl malonate ≥98%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
1,3-Propanedioic acid dimethyl ester, Malonic acid dimethyl ester
Purity: 98+%
CAS No.: 108-59-8
Molecular Formula: C5H8O4
Formula Weight: 132.12
Flash Point: 90°(194°F)
Refractive Index: 1.4130
Melting Point: -62°
Boiling Point: 180-181°
Beilstein Reference: 774261
Merck Reference: 14,6096
MDL No.: MFCD00008460
Vzorec:
C₅H₈O₄ MW: 132,12 g/mol Bod varu: 180…181 °C Bod tání: –62 °C Hustota: 1,156 Bod vzplanutí: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008460 CAS číslo: 108-59-8 EINECS: 203-597-8 Merck Index: 14,6096 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...