Dimethyl malonate 99+%
Dodavatel: Thermo Scientific
Synonyms:
1,3-Propanedioic acid dimethyl ester, Malonic acid dimethyl ester
220700010
220702500
220700051
220700010.EA
4120
CZK
220700010.
220702500.
220700051.
Dimethyl malonate 99+%
Dimethyl malonate
Vzorec:
C₅H₈O₄ MW: 132,12 g/mol Bod varu: 180…181 °C (760 mmHg) Bod tání: –62 °C Hustota: 1,156 g/cm³ Bod vzplanutí: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008460 CAS číslo: 108-59-8 EINECS: 203-597-8 Merck Index: 12,06169 |
Výsledky specifikačního testu
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4120 to 1.4140 (20°C, 589 nm) |
Water | ≤0.1 % (K.F.) |
GC | ≥99.0 % |
Free acid | ≤0.1 % (as malonic acid) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...