Dietylmalonát ≥99%
Dodavatel: Thermo Fisher Scientific
Varování
|
Synonyms:
1,3-Propanedioic acid diethyl ester, Malonic acid diethyl ester
A15468.0C
A15468.0E
A15468.36
A15468.0CEA
13390
CZK
A15468.0C
A15468.0E
A15468.36
Dietylmalonát ≥99%
Dietylmalonát
Purity: 99%
CAS No.: 105-53-3
Molecular Formula: C7H12O4
Formula Weight: 160.17
Flash Point: 90°(194°F)
Physical Form: Liquid
Density: 1.055
Refractive Index: 1.4140
Odour: Pleasant
Melting Point: -50°
Boiling Point: 198-200°
Beilstein Reference: 774687
Merck Reference: 14,3823
MDL No.: MFCD00009195
Vzorec:
C₇H₁₂O₄ Bod varu: 198…200 °C Bod tání: –50 °C Hustota: 1,055 Bod vzplanutí: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009195 CAS číslo: 105-53-3 EINECS: 203-305-9 Merck Index: 14,3823 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...