Methyl-3-mercaptopropionate ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B20718.14
B20718.22
B20718.36
B20718.14EA
1060
CZK
B20718.14
B20718.22
B20718.36
Methyl-3-mercaptopropionate ≥98%
Methyl-3-mercaptopropionate
Purity: 98%
CAS No.: 2935-90-2
Molecular Formula: HSCH2CH2CO2CH3
Formula Weight: 120.17
Flash Point: 60°(140°F)
Refractive Index: 1.4640
Boiling Point: 164°
Beilstein Reference: 1742952
UN No.: 1228
MDL No.: MFCD00004895
Vzorec:
C₄H₈O₂S MW: 120,17 g/mol Bod varu: 164 °C Bod tání: –60 °C Hustota: 1,085 Bod vzplanutí: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004895 CAS číslo: 2935-90-2 EINECS: 220-912-4 UN: 1228 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...