Dimethyl carbonate ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A13104.0E
A13104.22
A13104.36
AB116181.2500
A13104.0EEA
3810
CZK
A13104.0E
A13104.22
A13104.36
ABCRAB116181.2500
Dimethyl carbonate ≥99%
Dimethyl carbonate
Purity: 99%
CAS No.: 616-38-6
Molecular Formula: C3H6O3
Formula Weight: 90.08
Flash Point: 16°(61°F)
Physical Form: Liquid
Density: 1.070
Refractive Index: 1.3680
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Pleasant
Melting Point: ca 4°
Boiling Point: 90°
Solubility: Insoluble in water. Miscible with alcohol, ether
Beilstein Reference: 635821
Merck Reference: 14,3241
UN No.: 1161
MDL No.: MFCD00008420
Vzorec:
C₃H₆O₃ MW: 90.08 Bod varu: 90 °C Bod tání: ∼ 4 °C Hustota: 1.07 Bod vzplanutí: 16 °C (61 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008420 CAS číslo: 616-38-6 EINECS: 210-478-4 UN: 1161 ADR: 3 II Merck Index: 12,06117 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...