Tisk…

Methyl myristate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tetradecylic acid methyl ester, Myristic acid methyl ester, Tetradecanoic acid methyl ester, Methyl tetradecanoate

M-Clarity™ quality level = MQ200

M3378-1G M3378-25G
SIALM3378-1GEA 1200 CZK
SIALM3378-1G SIALM3378-25G
Methyl myristate, Sigma-Aldrich®
Methyl myristate
Vzorec: C₁₅H₃₀O₂
MW: 242,4 g/mol
Bod varu: 323 °C (1013 hPa)
Bod tání: 18 °C
Hustota: 0,86715 g/cm³ (20 °C)
Bod vzplanutí: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008983
CAS číslo: 124-10-7
EINECS: 204-680-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR