Ethyl myristate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Tetradecanoic acid ethyl ester, Tetradecylic acid ethyl ester, Ethyl tetradecanoate, Myristic acid ethyl ester
M-Clarity™ quality level = MQ300
W244503-1KG-K
W244503-20KG-K
W244503-4KG-K
W244503-9KG-K
SIALW244503-1KG-KEA
4440
CZK
SIALW244503-1KG-K
SIALW244503-20KG-K
SIALW244503-4KG-K
SIALW244503-9KG-K
Ethyl myristate, Sigma-Aldrich®
Ethyl myristate
Vzorec:
C₁₆H₃₂O₂ MW: 256,43 g/mol Hustota: 0,86 g/cm³ (20 °C) |
MDL Number:
MFCD00008984 CAS číslo: 124-06-1 EINECS: 204-675-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...