Methyl propionate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ300
W274208-10KG-K
W274208-1KG-K
W274208-20KG-K
SIALW274208-10KG-KEA
22720
CZK
SIALW274208-10KG-K
SIALW274208-1KG-K
SIALW274208-20KG-K
Methyl propionate, Sigma-Aldrich®
Methyl propionate
Vzorec:
C₄H₈O₂ Bod varu: 79,7 °C (1013 hPa) Bod tání: –87 °C Hustota: 0,91 g/cm³ (20 °C) Bod vzplanutí: –2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009306 CAS číslo: 554-12-1 EINECS: 209-060-4 UN: 1248 ADR: 3,II Merck Index: 12,06191 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...