Methyl propionate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Propanoic acid methyl ester
M-Clarity™ quality level = MQ100
109258-100ML
109258-500ML
SIAL109258-100MLEA
1270
CZK
SIAL109258-100ML
SIAL109258-500ML
Methyl propionate, Sigma-Aldrich®
Methyl propionate
Vzorec:
C₄H₈O₂ MW: 88,11 g/mol Bod varu: 79,7 °C (1013 hPa) Bod tání: –87 °C Hustota: 0,91 g/cm³ (20 °C) Bod vzplanutí: –2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009306 CAS číslo: 554-12-1 EINECS: 209-060-4 UN: 1248 ADR: 3,II Merck Index: 12,06191 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...