ethylpropionát ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 99%
CAS No.: 105-37-3
Molecular Formula: CH3CH2CO2CH2CH3
Formula Weight: 102.13
Flash Point: 12°(54°F)
Refractive Index: 1.3850
Melting Point: -74°
Boiling Point: 99°
Beilstein Reference: 506287
Merck Reference: 14,3847
UN No.: 1195
MDL No.: MFCD00009308
Vzorec:
C₅H₁₀O₂ Bod varu: 99 °C Bod tání: –74 °C Hustota: 0,891 Bod vzplanutí: 12 °C (54 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009308 CAS číslo: 105-37-3 EINECS: 203-291-4 UN: 1195 ADR: 3,II Merck Index: 14,3847 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...