Tisk…

Butyl propionate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Propionic acid butyl ester

B20125.22 B20125.36
B20125.22EA 975 CZK
B20125.22 B20125.36
Butyl propionate ≥99%
Butyl propionate

Purity: 99% CAS No.: 590-01-2 Molecular Formula: CH3CH2CO2(CH2)3CH3 Formula Weight: 130.19 Flash Point: 38°(101°F) Refractive Index: 1.4020 Boiling Point: 144-146° Merck Reference: 14,1587 UN No.: 3272 MDL No.: MFCD00009448

Vzorec: C₇H₁₄O₂
MW: 130.19
Bod varu: 144…146 °C
Bod tání: –90 °C
Hustota: 0.87
Bod vzplanutí: 38 °C (101 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009448
CAS číslo: 590-01-2
EINECS: 209-669-5
UN: 3272
Merck Index: 12,01623

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR