Tisk…

Ethyl valerate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl Pentanoate, Valeric acid ethyl ester

M-Clarity™ quality level = MQ300

W246204-1KG-K W246204-20KG-K W246204-9KG-K
SIALW246204-1KG-KEA 1930 CZK
SIALW246204-1KG-K SIALW246204-20KG-K SIALW246204-9KG-K
Ethyl valerate, Sigma-Aldrich®
Ethyl valerate
Vzorec: C₇H₁₄O₂
MW: 130.19
Bod varu: 145
Bod tání: –91 °C
Hustota: 0.87 g/cm³ (20 °C)
Bod vzplanutí: 35
Storage Temperature: Ambient
MDL Number: MFCD00009479
CAS číslo: 539-82-2
EINECS: 208-726-1
UN: 3272
ADR: 3 III
Merck Index: 12,10042

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR