Tisk…

Ethyl valerate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Ethyl Pentanoate, Valeric acid ethyl ester

A14854.0F A14854.AE A14854.AP
A14854.0FEA 6660 CZK
A14854.0F A14854.AE A14854.AP
Ethyl valerate ≥98%
Ethyl valerate

Purity: 98% CAS No.: 539-82-2 Molecular Formula: CH3(CH2)3CO2CH2CH3 Formula Weight: 130.19 Flash Point: 38°(100°F) Refractive Index: 1.400 Melting Point: -92 to -90° Boiling Point: 142-145° Beilstein Reference: 1744680 Merck Reference: 14,9904 UN No.: 3272 MDL No.: MFCD00009479

Vzorec: C₇H₁₄O₂
MW: 130,19 g/mol
Bod varu: 142…145 °C
Bod tání: –92 to –90 °C
Hustota: 0,874
Bod vzplanutí: 38 °C (100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009479
CAS číslo: 539-82-2
EINECS: 208-726-1
UN: 3272
ADR: 3,III
Merck Index: 14,9904

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR