Methyl octanoate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate
M-Clarity™ quality level = MQ100
260673-25G
260673-100G
SIAL260673-25GEA
1520
CZK
SIAL260673-25G
SIAL260673-100G
Methyl octanoate, Sigma-Aldrich®
Methyl octanoate
Vzorec:
C₉H₁₈O₂ MW: 158,24 g/mol Bod varu: 190 °C (1013 hPa) Bod tání: –36,7 °C Hustota: 0,877 g/cm³ (20 °C) Bod vzplanutí: 68 °C |
MDL Number:
MFCD00009551 CAS číslo: 111-11-5 EINECS: 203-835-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...