Tisk…

Methyl octanoate ≥98%

Dodavatel: Thermo Scientific

Synonyms: Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate

A10991.18 A10991.30
A10991.18EA 1770 CZK
A10991.18 A10991.30
Methyl octanoate ≥98%
Methyl octanoate

Purity: 99% CAS No.: 111-11-5 Molecular Formula: CH3(CH2)6CO2CH3 Formula Weight: 158.24 Flash Point: 72°(161°F) Refractive Index: 1.4170 Melting Point: -40° Boiling Point: 193-194° Beilstein Reference: 1752270 MDL No.: MFCD00009551

Vzorec: C₉H₁₈O₂
MW: 158,24 g/mol
Bod varu: 193…194 °C
Bod tání: –40 °C
Hustota: 0,875
Bod vzplanutí: 72 °C (161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009551
CAS číslo: 111-11-5
EINECS: 203-835-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR