Methyl octanoate 99%
Dodavatel: Thermo Scientific
Synonyms:
Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate
414620250
414621000
414625000
ACRO414620250EA
1880
CZK
ACRO414620250
ACRO414621000
ACRO414625000
Methyl octanoate 99%
Methyl octanoate
Vzorec:
C₉H₁₈O₂ MW: 158,24 g/mol Bod varu: 79 °C (12 mmHg) Bod tání: –36,7 °C Hustota: 0,878 g/cm³ Bod vzplanutí: 73 °C |
MDL Number:
MFCD00009551 CAS číslo: 111-11-5 EINECS: 203-835-0 |
Výsledky specifikačního testu
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4165 to 1.4185 (20°C, 589 nm) |
GC | ≥98.5 % |
Specific gravity | (20°C) 0.868 to 0.888 |
Acid value | ≤1 mg KOH/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...