Ethyl octanoate ≥99%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
Caprylic acid ethyl ester, Octanoic acid ethyl ester, Ethyl caprylate
B23450.22
B23450.36
B23450.22EA
845
CZK
B23450.22
B23450.36
Ethyl octanoate ≥99%
Ethyl octanoate
Purity: 99%
CAS No.: 106-32-1
Molecular Formula: C10H20O2
Formula Weight: 172.27
Flash Point: 79°(174°F)
Refractive Index: 1.4170
Odour: Ester-like
Melting Point: -47°
Boiling Point: 206-208°
Merck Reference: 14,3778
MDL No.: MFCD00009552
Vzorec:
C₁₀H₂₀O₂ MW: 172,27 g/mol Bod varu: 206…208 °C Bod tání: –47 °C Hustota: 0,872 Bod vzplanutí: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009552 CAS číslo: 106-32-1 EINECS: 203-385-5 Merck Index: 14,3778 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...