Tisk…

Ethyl octanoate 99+%

Dodavatel: Thermo Scientific

Synonyms: Caprylic acid ethyl ester, Octanoic acid ethyl ester, Ethyl caprylate

150510050 150511000 150515000
ACRO150510050EA 857 CZK
ACRO150510050 ACRO150511000 ACRO150515000
Ethyl octanoate 99+%
Ethyl octanoate
Vzorec: C₁₀H₂₀O₂
MW: 172,27 g/mol
Bod varu: 206 °C (1013 hPa)
Bod tání: –47 °C
Hustota: 0,868 g/cm³
Bod vzplanutí: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00009552
CAS číslo: 106-32-1
EINECS: 203-385-5
Merck Index: 13,03813

Objednat

Výsledky specifikačního testu

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4200 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥99.0 %
Specific gravity (20°C) 0.866 to 0.87
Acid value ≤1 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR