Tisk…

Ethyl stearate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Stearic acid ethyl ester, Ethyl octadecanoate

M-Clarity™ quality level = MQ200

S8269-5G S8269-25G
SIALS8269-5GEA 2840 CZK
SIALS8269-5G SIALS8269-25G
Ethyl stearate, Sigma-Aldrich®
Ethyl stearate
Vzorec: C₂₀H₄₀O₂
MW: 312,54 g/mol
Bod varu: 365 °C (1013 hPa)
Bod tání: 31 °C
Hustota: 0,8973 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009006
CAS číslo: 111-61-5
EINECS: 203-887-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR