Tisk…

Propyl formate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Formic acid propyl ester

M-Clarity™ quality level = MQ300

W294306-1KG-K W294306-4KG-K
SIALW294306-1KG-KEA 0 CZK
SIALW294306-1KG-K SIALW294306-4KG-K
Propyl formate, Sigma-Aldrich®
Propyl formate
Vzorec: C₄H₈O₂
MW: 88,11 g/mol
Bod varu: 81 °C (1013 hPa)
Bod tání: –93 °C
Hustota: 0,906 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003295
CAS číslo: 110-74-7
EINECS: 203-798-0
UN: 1281
ADR: 3,II
Merck Index: 12,08043

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR