Tisk…

Propyl formate ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Formic acid propyl ester

A18090.AE A18090.AP
A18090.AEEA 1430 CZK
A18090.AE A18090.AP
Propyl formate ≥97%
Propyl formate

Purity: 97% CAS No.: 110-74-7 Molecular Formula: HCO2CH2CH2CH3 Formula Weight: 88.11 Flash Point: -4°(25°F) Refractive Index: 1.3750 Melting Point: -93° Boiling Point: 80-81° Beilstein Reference: 1739272 Merck Reference: 14,7858 UN No.: 1281 MDL No.: MFCD00003295

Vzorec: C₄H₈O₂
MW: 88,11 g/mol
Bod varu: 80…81 °C
Bod tání: –93 °C
Hustota: 0,904
Bod vzplanutí: –4 °C (25 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003295
CAS číslo: 110-74-7
EINECS: 203-798-0
UN: 1281
ADR: 3,II
Merck Index: 14,7858

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR