Tisk…

Ethyl hexanoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl caproate, Caproic acid ethyl ester, Hexanoic acid ethyl ester

M-Clarity™ quality level = MQ300

W243906-1KG-K W243906-20KG-K W243906-4KG-K W243906-9KG-K
SIALW243906-1KG-KEA 2900 CZK
SIALW243906-1KG-K SIALW243906-20KG-K SIALW243906-4KG-K SIALW243906-9KG-K
Ethyl hexanoate, Sigma-Aldrich®
Ethyl hexanoate
Vzorec: C₈H₁₆O₂
MW: 144,21 g/mol
Bod varu: 168 °C (1013 hPa)
Bod tání: –67 °C
Hustota: 0,87 g/cm³ (20 °C)
Bod vzplanutí: 52 °C
MDL Number: MFCD00009511
CAS číslo: 123-66-0
EINECS: 204-640-3
UN: 3272
ADR: 3,III
Merck Index: 13,03812

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR