Ethyl hexanoate ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ethyl caproate, Caproic acid ethyl ester, Hexanoic acid ethyl ester
A13985.0E
A13985.22
A13985.36
A13985.0EEA
8000
CZK
A13985.0E
A13985.22
A13985.36
Ethyl hexanoate ≥99%
Ethyl hexanoate
Purity: 99%
CAS No.: 123-66-0
Molecular Formula: CH3(CH2)4CO2CH2CH3
Formula Weight: 144.21
Flash Point: 49°(121°F)
Refractive Index: 1.4075
Melting Point: -67°
Boiling Point: 166°
Merck Reference: 14,3777
UN No.: 3272
MDL No.: MFCD00009511
Vzorec:
C₈H₁₆O₂ MW: 144,21 g/mol Bod varu: 166 °C Bod tání: –67 °C Hustota: 0,873 Bod vzplanutí: 49 °C (121 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009511 CAS číslo: 123-66-0 EINECS: 204-640-3 UN: 3272 ADR: 3,III Merck Index: 14,3777 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...