Tisk…

Octyl acetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Acetic acid octyl ester

M-Clarity™ quality level = MQ300

W280607-1KG-K W280607-20KG-K W280607-4KG-K
SIALW280607-1KG-KEA 3460 CZK
SIALW280607-1KG-K SIALW280607-20KG-K SIALW280607-4KG-K
Octyl acetate, Sigma-Aldrich®
Octyl acetate
Vzorec: C₁₀H₂₀O₂
MW: 172,27 g/mol
Bod varu: 195…199 °C (1013 hPa)
Bod tání: –38,5 °C
Hustota: 0,885 g/cm³ (0 °C)
MDL Number: MFCD00009562
CAS číslo: 112-14-1
EINECS: 203-939-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR