Tisk…

Hexyl acetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Acetic acid hexyl ester

M-Clarity™ quality level = MQ300

W256528-100G-K W256528-1KG-K W256528-4KG-K
SIALW256528-100G-KEA 2730 CZK
SIALW256528-100G-K SIALW256528-1KG-K SIALW256528-4KG-K
Hexyl acetate, Sigma-Aldrich®
Hexyl acetate
Vzorec: C₈H₁₆O₂
MW: 144,21 g/mol
Bod varu: 338 °C (1013 hPa)
Bod tání: –80 °C
Hustota: 0,87 g/cm³ (20 °C)
Bod vzplanutí: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009524
CAS číslo: 142-92-7
EINECS: 205-572-7
UN: 3271
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR