Hexyl acetate, Sigma-Aldrich®
Dodavatel: Merck
|
|
M-Clarity™ quality level = MQ200
108154-1L
108154-25ML
108154-500ML
SIAL108154-1LEA
4520
CZK
SIAL108154-1L
SIAL108154-25ML
SIAL108154-500ML
Hexyl acetate, Sigma-Aldrich®
Hexyl acetate
Vzorec:
C₈H₁₆O₂ Bod varu: 338 °C (1013 hPa) Bod tání: –80 °C Hustota: 0,87 g/cm³ (20 °C) Bod vzplanutí: 56 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009524 CAS číslo: 142-92-7 EINECS: 205-572-7 UN: 3271 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...