Tisk…
Decyl acetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Acetic acid decyl ester
M-Clarity™ quality level = MQ300
W236705-1KG-K W236705-4KG-K W236705-9KG-K
SIALW236705-1KG-KEA 4950 CZK
SIALW236705-1KG-K SIALW236705-4KG-K SIALW236705-9KG-K
Decyl acetate, Sigma-Aldrich®
Decyl acetate
Vzorec: C₁₂H₂₄O₂
MW: 200.32
MDL Number: MFCD00026538
CAS číslo: 112-17-4
EINECS: 203-942-2