Tisk…

Methyl laurate ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester

A12492.06 A12492.14 A12492.22
A12492.06EA 1450 CZK
A12492.06 A12492.14 A12492.22
Methyl laurate ≥99%
Methyl laurate

Purity: 99% CAS No.: 111-82-0 Molecular Formula: CH3(CH2)10CO2CH3 Formula Weight: 214.35 Flash Point: 125°(257°F) Refractive Index: 1.4325 Melting Point: 4-5° Boiling Point: 261-262° Beilstein Reference: 1767780 MDL No.: MFCD00008966

Vzorec: C₁₃H₂₆O₂
Bod varu: 261…262 °C
Bod tání: 4…5 °C
Hustota: 0,870
Bod vzplanutí: 125 °C (257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008966
CAS číslo: 111-82-0
EINECS: 203-911-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR