Methylbutyrát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Butyric acid methyl ester, Butanoic acid methyl ester
M-Clarity™ quality level = MQ300
W269301-1KG-K
W269301-20KG-K
W269301-5KG-K
SIALW269301-1KG-KEA
3240
CZK
SIALW269301-1KG-K
SIALW269301-20KG-K
SIALW269301-5KG-K
Methylbutyrát, Sigma-Aldrich®
Methylbutyrát
Vzorec:
C₅H₁₀O₂ MW: 102.13 Bod varu: 102 Bod tání: –84 °C Hustota: 0.8984 g/cm³ (20 °C) Bod vzplanutí: 14 |
MDL Number:
MFCD00009391 CAS číslo: 623-42-7 EINECS: 210-792-1 UN: 1237 ADR: 3 II Merck Index: 13,06062 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...