Methylbutyrát 98+%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Butyric acid methyl ester, Butanoic acid methyl ester
L04250.AE
L04250.AP
L04250.AEEA
810
CZK
L04250.AE
L04250.AP
Methylbutyrát 98+%
Methylbutyrát
Purity: 98+%
CAS No.: 623-42-7
Molecular Formula: CH3CH2CH2COOCH3
Formula Weight: 102.14
Flash Point: 11°(51°F)
Refractive Index: 1.3860
Melting Point: -85 to -84°
Boiling Point: 100-102°
Beilstein Reference: 1740743
Merck Reference: 14,6035
UN No.: 1237
MDL No.: MFCD00009391
Vzorec:
C₅H₁₀O₂ MW: 102.13 Bod varu: 100…102 °C Bod tání: –85…–84 °C Hustota: 0.8984 Bod vzplanutí: 11 °C (51 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009391 CAS číslo: 623-42-7 EINECS: 210-792-1 UN: 1237 ADR: 3 II Merck Index: 13,06062 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...