Tisk…

ethylbutyrát, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Butanoic acid ethyl ester, Butyric acid ethyl ester

M-Clarity™ quality level = MQ100

E15701-500ML E15701-1L E15701-25ML
SIALE15701-500MLEA 985 CZK
SIALE15701-500ML SIALE15701-1L SIALE15701-25ML
ethylbutyrát, Sigma-Aldrich®
ethylbutyrát
Vzorec: C₆H₁₂O₂
MW: 116.16
Bod varu: 252
Bod tání: –92 °C
Hustota: 0.88
Bod vzplanutí: 26
Storage Temperature: Ambient
MDL Number: MFCD00009394
CAS číslo: 105-54-4
EINECS: 203-306-4
UN: 1180
ADR: 3 III
Merck Index: 13,03810

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR