Paracetamol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
M-Clarity™ quality level = MQ200
A5000-100G
A5000-1KG
A5000-500G
SIALA5000-100GEA
1490
CZK
SIALA5000-100G
SIALA5000-1KG
SIALA5000-500G
Paracetamol, Sigma-Aldrich®
Paracetamol
Vzorec:
C₈H₉NO₂ MW: 151,16 g/mol Bod varu: 388 °C (1013 hPa) Bod tání: 168 °C Hustota: 1,34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002328 CAS číslo: 103-90-2 EINECS: 203-157-5 Merck Index: 13,00048 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...