Metacetamol ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
N-(3-Hydroxyphenyl)acetamide, N-Acetyl-3-aminophenol, Paracetamol, 3-Acetamidophenol, 3'-Hydroxyacetanilide
A12764.14
A12764.22
A12764.14EA
1110
CZK
A12764.14
A12764.22
Metacetamol ≥99%
Metacetamol
Purity: 99%
CAS No.: 621-42-1
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 146-149°
Beilstein Reference: 907998
Vzorec:
C₈H₉NO₂ MW: 151,16 g/mol Bod varu: 351 °C (1013 hPa) Bod tání: 146…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002263 CAS číslo: 621-42-1 EINECS: 210-687-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...