Tisk…

2'-Aminoacetanilide 98%

Dodavatel: Thermo Scientific
Varování

H27343.14 H27343.06
H27343.14EA 6520 CZK
H27343.14 H27343.06
2'-Aminoacetanilide 98%
2'-Aminoacetanilide

Purity: 98% CAS No.: 34801-09-7 Molecular Formula: C8H10N2O Formula Weight: 150.18 Melting Point: 133-137° MDL No.: MFCD00210388

Vzorec: C₈H₁₀N₂O
MW: 150,18 g/mol
Bod tání: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00210388
CAS číslo: 34801-09-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR