4'-Methylacetanilide ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
p-Acetotoluidid, p-Acetotoluidide, N-(4-Methylphenyl)acetamide
A13721.14
A13721.22
A13721.36
A13721.14EA
876
CZK
A13721.14
A13721.22
A13721.36
4'-Methylacetanilide ≥98%
4'-Methylacetanilide
Purity: 98+%
CAS No.: 103-89-9
Molecular Formula: C9H11NO
Formula Weight: 149.19
Melting Point: 147-150°
Boiling Point: 306-308°
Merck Reference: 14,74
MDL No.: MFCD00008677
Vzorec:
C₉H₁₁NO MW: 149,19 g/mol Bod varu: 306…308 °C Bod tání: 147…150 °C Hustota: 1,22 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008677 CAS číslo: 103-89-9 EINECS: 203-155-4 Merck Index: 14,74 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...