ACES, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid
MQ200
A3594-100G
A3594-1KG
A3594-500G
SIALA3594-100GEA
15730
CZK
SIALA3594-100G
SIALA3594-1KG
SIALA3594-500G
ACES, Sigma-Aldrich®
ACES
Vzorec:
H₂NCOCH₂NHCH₂CH₂SO₃H MW: 182.2 Bod tání: 270…295 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008030 CAS číslo: 7365-82-4 EINECS: 230-908-4 Merck Index: 13,00037 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...